Learn More
Medchemexpress LLC HY-15437 5mg Medchemexpress, SL327 CAS:305350-87-2 Purity:>98%
SDP
Catalog No. 502030625
Encompass_Preferred
Medchemexpress, HY-15437 5mg SL327 CAS:305350-87-2 SL327 inhibits MEK1 and MEK2, with IC50 values of 180 nM and 220 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0625 Supplier Medchemexpress LLC Supplier No. HY154375MG
Medchemexpress, HY-15437 5mg SL327 CAS:305350-87-2 SL327 inhibits MEK1 and MEK2, with IC50 values of 180 nM and 220 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.