Learn More
Aviva Systems Biology Corp SLA Antibody - middle region (ARP42317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLA. Alternate names: SLA1, SLAPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58180