Learn More
Aviva Systems Biology Corp SLA Antibody - N-terminal region (ARP41159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLA/LP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLA/LP. Alternate names: LP, SLA, PCH2D, SLA/LPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57744