Learn More
Aviva Systems Biology Corp SLA2 Antibody - N-terminal region (ARP60508_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60508_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SLA2. Alternate names: C20orf156, FLJ21992, MGC49845, SLAP-2, SLAP2, MARSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-066