Learn More
Mopec Slab and Roller Cleaner
SDP
Catalog No. 22446315 Shop All Mopec Products
Change view
Click to view available options
Quantity:
6 oz.
10 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-446-315 6 oz.
22-446-312 10 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-315 Supplier Mopec Supplier No. FS367
Add to Cart
Add to Cart
This fast-acting cleaning solution removes inks, grease, buildup, and tough residues from printing slabs and rollers. Its effective formula supports routine equipment cleaning while helping maintain performance and longevity by reducing accumulated residue on working surfaces.
Construction
  • Aerosol spray can
  • Fast-acting ink solvent
Performance
  • Loosens and removes fingerprint ink from rubber rollers, glass or stainless steel slabs
Specifications
Product Type Fingerprint Slab Cleaner
Quantity 6 oz.