Learn More
Aviva Systems Biology Corp SLAIN1 Antibody - middle region (ARP52687_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52687_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLAIN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLAIN1. Alternate names: C13orf32, FLJ30046, MGC131899ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60803