Learn More
ACROBiosystems Recombinant Protein;Human SLAMF1 / SLAM / CD150 Protein;HEK293;1MG;SL1,SLAMF1,CD150,CDw150,SLAM,IPO-3
Catalog No. 501657985
Encompass_Preferred
Recombinant Protein;Human SLAMF1 / SLAM / CD150 Protein;1MG;Human SLAMF1 (SL1-H52H0) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7985 Supplier ACROBiosystems Supplier No. SL1H52H01MG
Recombinant Protein;Human SLAMF1 / SLAM / CD150 Protein;1MG;Human SLAMF1 (SL1-H52H0) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.