Learn More
Aviva Systems Biology Corp SLAMF6 Antibody - N-terminal region (ARP50224_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50224_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLAMF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLAMF6. Alternate names: KALI, KALIb, Ly108, MGC104953, NTB-A, NTBA, SF2000, CD352ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60291