Learn More
Sino Biological SLAMF7 / CD319 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11691MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SLAMF7 / CD319 (rh SLAMF7 / CD319; Catalog#11691-H08H; NP_067004.3; Met1-Met226). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8653