Learn More
Genscript Corporation SLAMF7/CRACC/CD319Biotin Hi
Supplier:  Genscript Corporation Z04616500
Encompass
CD2-like receptor activating cytotoxic cells (CRACC) also known as CS1 novel Ly9 SLAMF7 and CD319 is a 65-75 kDa type I transmembrane glycoprotein in the SLAM subgroup of the CD2 family Self-ligand receptor of the signaling lymphocytic activation molecule (SLAM) family SLAM receptors triggered by homo- or heterotypic cell-cell interactions are modulating the activation and differentiation of a wide variety of immune cells and thus are involved in the regulation and interconnection of both innate and adaptive immune response
Catalog No. 50-000-57607