Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human SLAMF7 / CRACC / CD319 Protein, His Tag
Catalog No. 501956961
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human SLAMF7, His Tag (Cat. No. SL7-HF2H7) is expressed from human HEK293 cells. It contains AA Ser 23 - Met 226 (Accession # AAH27867). It is the FITC labeled form of Human SLAMF7, His Tag (Cat. No. SL7-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6961 Supplier ACROBiosystems Supplier No. SL7HF2H725UG
Recombinant Protein;25UG;FITC-Labeled Human SLAMF7, His Tag (Cat. No. SL7-HF2H7) is expressed from human HEK293 cells. It contains AA Ser 23 - Met 226 (Accession # AAH27867). It is the FITC labeled form of Human SLAMF7, His Tag (Cat. No. SL7-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.