Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque SLAMF7 / CRACC / CD319 Protein;HEK293;1MG;SL7,SLAMF7,CD319,CS1,CRACC,19A,FOAP-12,SLAMF7,FOAP12
Supplier:  ACROBiosystems SL7R52H71MG
Encompass_Preferred
Recombinant Protein;Rhesus macaque SLAMF7 / CRACC / CD319 Protein;1MG;Rhesus macaque SLAMF7 (SL7-R52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # F7HQ72-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 35-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7799