Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human SLAMF7 / CRACC / CD319 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;SL7,SLAMF7,CD319,CS1,CRACC,19A,FOAP-12,SLAMF7,FOAP12
Catalog No. 501658301
Encompass_Preferred
Recombinant Protein;Biotinylated Human SLAMF7 / CRACC / CD319 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human SLAMF7, His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8301 Supplier ACROBiosystems Supplier No. SL7H82E025UG
Recombinant Protein;Biotinylated Human SLAMF7 / CRACC / CD319 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human SLAMF7, His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.