Learn More
Aviva Systems Biology Corp Slc10a2 Antibody - N-terminal region (ARP43749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc10a2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: ASBT, ISBTApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-096