Learn More
Aviva Systems Biology Corp SLC10A3 Antibody - middle region (ARP49461_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49461_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC10A3. It was validated on Western Blot. SLC10A3,solute carrier family 10 (sodium/bile acid cotransporter family), member 3,P3; DXS253E; SLC10A3,P3 protein,P3 protein,Protein P3,OTTHUMP00000015406,solute carrier family 10 member 3The immunogen is a synthetic peptide directed towards the middle region of human SLC10A3. Alternate names: DXS253E, P3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-423