Learn More
Aviva Systems Biology Corp SLC10A4 Antibody - N-terminal region (ARP44121_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44121_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC10A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC10A4. Alternate names: MGC29802, P4ApplicationWB. Species reactivity: Cow, Human, Pig, Rabbit
Catalog No. 50-014-58724