Learn More
Aviva Systems Biology Corp SLC10A5 Antibody - C-terminal region (ARP43773_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43773_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC10A5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC10A5. Alternate names: P5ApplicationIHC, WB. Species reactivity: Horse, Human, Pig, Rat
Catalog No. 50-014-53374