Learn More
Aviva Systems Biology Corp SLC12A1 Antibody - N-terminal region (ARP41388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC12A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC12A1. Alternate names: BSC1, MGC48843, NKCC2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53296