Learn More
Aviva Systems Biology Corp SLC12A2 Antibody - C-terminal region (ARP43805_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43805_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC12A2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC12A2. Alternate names: BSC, BSC2, MGC104233, NKCC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53376