Learn More
Biomatik Corporation Slc12A2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11244
Encompass_Preferred
Slc12A2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC12A2. The Slc12A2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Family 12 Member 2 Protein (110-282Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC12A2. Accession Number: P55011. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9247