Learn More
Aviva Systems Biology Corp SLC12A3 Antibody - middle region (ARP43742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC12A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC12A3. Alternate names: NCCT, TSCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58614