Learn More
Aviva Systems Biology Corp Slc12a5 Antibody - N-terminal region (ARP44012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc12a5. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: KCC2, mKIAA1176ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-117