Learn More
Aviva Systems Biology Corp SLC12A8 Antibody - N-terminal region (ARP44039_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44039_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC12A8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC12A8. Alternate names: CCC9, DKFZp686L18248, FLJ23188ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58697