Learn More
Aviva Systems Biology Corp SLC13A3 Antibody - middle region (ARP41439_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41439_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC13A3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC13A3. Alternate names: NADC3, SDCT2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57855