Learn More
Aviva Systems Biology Corp SLC14A1 Antibody - C-terminal region (ARP48116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC14A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC14A1. Alternate names: FLJ33745, FLJ41687, HUT11, HsT1341, JK, RACH1, UT-B1, UT1, UTEApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Sheep
Catalog No. 50-014-59718