Learn More
Aviva Systems Biology Corp Slc15a3 Antibody - C-terminal region (ARP43965_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43965_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc15a3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Ci1, cI-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-115