Learn More
Aviva Systems Biology Corp SLC15A4 Antibody - middle region (ARP44109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC15A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC15A4. Alternate names: FP12591, PHT1, PTR4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58720