Learn More
Aviva Systems Biology Corp SLC16A1 Antibody - middle region (ARP43841_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43841_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC16A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC16A1. Alternate names: FLJ36745, MCT, MCT1, MGC44475, HHF7ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58642