Learn More
Aviva Systems Biology Corp SLC16A12 Antibody - N-terminal region (ARP44186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC16A12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC16A12. Alternate names: DKFZp686E188, MCT12, CJMGApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58741