Learn More
Aviva Systems Biology Corp SLC17A2 Antibody - middle region (ARP33806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC17A2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC17A2. Alternate names: NPT3ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55153