Learn More
Aviva Systems Biology Corp SLC17A3 Antibody - middle region (ARP42305_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42305_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC17A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC17A3. Alternate names: NPT4ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-58177