Learn More
Aviva Systems Biology Corp SLC17A4 Antibody - middle region (ARP43906_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43906_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC17A4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC17A4. Alternate names: KAIA2138, KIAA2138, MGC129623ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53382