Learn More
Aviva Systems Biology Corp SLC17A5 Antibody - N-terminal region (ARP43932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC17A5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC17A5. Alternate names: AST, FLJ22227, FLJ23268, ISSD, NSD, SD, SIALIN, SIASD, SLDApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53386