Learn More
ABclonal Technology SLC17A6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15177500UL
Encompass_Preferred
Predicted to enable L-glutamate transmembrane transporter activity and neurotransmitter transmembrane transporter activity. Involved in neurotransmitter loading into synaptic vesicle. Predicted to be located in synaptic vesicle. Predicted to be active in excitatory synapse. Predicted to be integral component of synaptic vesicle membrane.
Catalog No. 50-001-07047