Learn More
Aviva Systems Biology Corp SLC18A1 Antibody - N-terminal region (ARP35650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC18A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC18A1. Alternate names: CGAT, VAT1, VMAT1ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-55805