Learn More
Aviva Systems Biology Corp SLC18A2 Antibody - N-terminal region (ARP43845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC18A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC18A2. Alternate names: MGC120477, MGC120478, MGC26538, SVAT, SVMT, VAT2, VMAT2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58644