Learn More
Aviva Systems Biology Corp SLC1A1 Antibody - N-terminal region (ARP43865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC1A1. Alternate names: EAAC1, EAAT3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58652