Learn More
Aviva Systems Biology Corp Slc1a4 Antibody - C-terminal region (ARP43828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc1a4. It was validated on Western Blot. . Alternate names: ASCT1, AW045657, SATT, ASCT-1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-101