Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG 40 Castor Oil, Spectrum™ Chemical
SDP

Catalog No. 18605002 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-002 4 L Steel Can
18-605-001 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-002 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P24044LT
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P2404, 61791-12-6

Spectrum™ Chemical PEG 40 Castor Oil is a vegetable oil formed from the seeds of the castor oil plant. It is used to produce lubricants, hydraulic and brake fluids, pharmaceuticals, and cold resistant plastics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 61791-12-6

Specifications

Acid Value 2
CAS Max % 1
Quantity 4 L
Moisture 0.03
Packaging Steel Can
Grade Reagent
Chemical Name or Material PEG 40 Castor Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.