Learn More
Medchemexpress LLC SLC1A5/ASCT2 Antibody (YA1180) | 10 UL
SDP
Catalog No. 5000441789
Encompass_Preferred
SLC1A5/ASCT2 Antibody (YA1180) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets SLC1A5/ASCT2. This antibody is intended for research use only and aids in the study of sodium-coupled antiporters of neutral amino acids, which are crucial for cellular transport processes.
  • Rabbit host
  • Recombinant, monoclonal clonality
  • Human species reactivity
  • Affinity purified
  • Non-conjugated
  • IgG isotype
  • Liquid appearance
  • IHC-P (Immunohistochemistry-Paraffin) application
  • Stored at -20°C for 1 year
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, and 0.05% BSA.
Catalog No. 50-004-41789 Supplier Medchemexpress LLC Supplier No. HYP8143510UL
SLC1A5/ASCT2 Antibody (YA1180) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets SLC1A5/ASCT2. This antibody is intended for research use only and aids in the study of sodium-coupled antiporters of neutral amino acids, which are crucial for cellular transport processes.
  • Rabbit host
  • Recombinant, monoclonal clonality
  • Human species reactivity
  • Affinity purified
  • Non-conjugated
  • IgG isotype
  • Liquid appearance
  • IHC-P (Immunohistochemistry-Paraffin) application
  • Stored at -20°C for 1 year
  • Formulated in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, and 0.05% BSA.