Learn More
ACROBiosystems ACROBiosystems Human SLC1A5 Protein, His Tag, TrxA Tag
Supplier:  ACROBiosystems SL5H51491MG
Encompass_Preferred
Recombinant Protein;1MG;Human SLC1A5, His Tag, TrxA Tag (SL5-H5149) is expressed from E.coli cells. It contains AA Gln 154 - Gln 224 (Accession # Q15758-1).;This protein carries a polyhistidine tag at the N-terminus, followed by a TrxA tag. The protein has a calculated MW of 22.5 kDa. The protein migrates as 25 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7175