Learn More
Aviva Systems Biology Corp SLC22A1 Antibody - C-terminal region (ARP41640_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41640_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC22A1. Alternate names: HOCT1, OCT1, oct1_cdsApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57930