Learn More
Aviva Systems Biology Corp SLC22A10 Antibody - middle region (ARP43790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC22A10. Alternate names: OAT5, hOAT5ApplicationWB. Species reactivity: Human
Catalog No. 50-014-63052