Learn More
Aviva Systems Biology Corp SLC22A11 Antibody - N-terminal region (ARP43993_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43993_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A11. Alternate names: MGC34282, OAT4, hOAT4ApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-014-53391