Learn More
Aviva Systems Biology Corp SLC22A13 Antibody - N-terminal region (ARP43878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A13. Alternate names: OCTL1, OCTL3, ORCTL3, OAT10, ORCTL-3ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58656