Learn More
Aviva Systems Biology Corp SLC22A14 Antibody - N-terminal region (ARP43892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A14. Alternate names: MGC163415, OCTL2, OCTL4, ORCTL4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse
Catalog No. 50-014-58660