Learn More
Aviva Systems Biology Corp SLC22A16 Antibody - N-terminal region (ARP44073_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44073_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A16. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A16. Alternate names: CT2, FLIPT2, OCT6, OKB1, dJ261K5.1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53401