Learn More
Aviva Systems Biology Corp SLC22A2 Antibody - N-terminal region (ARP43847_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43847_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A2. Alternate names: MGC32628, OCT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58645