Learn More
Aviva Systems Biology Corp Slc22a3 Antibody - middle region (ARP44027_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44027_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc22a3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: EMT, Oct3, Orct3, Slca22a3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-285-120