Learn More
Aviva Systems Biology Corp SLC22A3 Antibody - N-terminal region (ARP44026_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44026_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC22A3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC22A3. Alternate names: EMT, EMTH, OCT3ApplicationIHC, WB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-53394